Invitrogen
Manufacturer Code:PA563928
Catalog # PIPA563928
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human ASAH2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acylsphingosine deacylase 2; BCDase; BCDase HNAC1 LCDase N-CDase NCDase N-acylsphingosine amidohydrolase 2 acylsphingosine deacylase 2 mitochondrial ceramidase neutral ceramidase neutral/alkaline ceramidase non-lysosomal ceramidase; hCD; LCDase; mitochondrial ceramidase; N-acylsphingosine amidohydrolase (non-lysosomal ceramidase) 2; N-acylsphingosine amidohydrolase 2; N-CDase; Neutral ceramidase; Neutral ceramidase soluble form; neutral/alkaline ceramidase; Non-lysosomal ceramidase
Gene Aliases: ASAH2; BCDase; HNAC1; LCDase; N-CDase; NCDase
UniProt ID: (Human) Q9NR71
Entrez Gene ID: (Human) 56624
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.