Invitrogen
Manufacturer Code:PA548033
Catalog # PIPA548033
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombit mouse ASAH2/N-acylsphingosine Amidohydrolase-2 Thr34-Thr756 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: Acylsphingosine deacylase 2; BCDase HNAC1 LCDase N-CDase NCDase N-acylsphingosine amidohydrolase 2 acylsphingosine deacylase 2 mitochondrial ceramidase neutral ceramidase neutral/alkaline ceramidase non-lysosomal ceramidase; N-acylsphingosine amidohydrolase 2; N-CDase; NCDase; Neutral ceramidase; Neutral ceramidase soluble form; neutral/alkaline ceramidase
Gene Aliases: AI585898; Asah2
UniProt ID: (Mouse) Q9JHE3
Entrez Gene ID: (Mouse) 54447
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.