Invitrogen
Manufacturer Code:PA5117113
Catalog # PIPA5117113
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein of human ASAH2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acylsphingosine deacylase 2; Acylsphingosine deacylase 2 AI585898 ASAH2 BCDase hCD HNAC1 LCDase mitochondrial ceramidase N-acylsphingosine amidohydrolase (non-lysosomal ceramidase) 2 N-acylsphingosine amidohydrolase 2 NCDase N-CDase Neutral ceramidase Neutral ceramidase soluble form neutral/alkaline ceramidase non-lysosomal ceramidase; BCDase; hCD; LCDase; mitochondrial ceramidase; N-acylsphingosine amidohydrolase (non-lysosomal ceramidase) 2; N-acylsphingosine amidohydrolase 2; N-CDase; NCDase; Neutral ceramidase; Neutral ceramidase soluble form; neutral/alkaline ceramidase; Non-lysosomal ceramidase
Gene Aliases: AI585898; ASAH2; BCDase; HNAC1; LCDase; N-CDase; NCDase
UniProt ID: (Human) Q9NR71, (Mouse) Q9JHE3, (Rat) Q91XT9
Entrez Gene ID: (Human) 56624, (Mouse) 54447, (Rat) 114104
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.