Abnova
Manufacturer Code:PAB8552
Catalog # 89-133-745
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to residues surrounding amino acids 741 of human ASAH2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of ASAH2.
For Research Use Only
Protein Aliases: Acylsphingosine deacylase 2; BCDase; hCD; HNAC1; LCDase; MGC129777; mitochondrial ceramidase; N-acylsphingosine amidohydrolase (non-lysosomal ceramidase) 2; N-acylsphingosine amidohydrolase 2; N-CDase; NCDase; Neutral ceramidase; Neutral ceramidase soluble form; neutral/alkaline ceramidase; Non-lysosomal ceramidase
Gene Aliases: ASAH2; BCDase; HNAC1; LCDase; N-CDase; NCDase
UniProt ID: (Human) Q9NR71
Entrez Gene ID: (Human) 56624
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.