Abnova
Manufacturer Code:H00000427M01
Catalog # 89-016-220
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2C9 |
Immunogen | ASAH1 (NP_808592 25 a.a. ~ 124 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant ASAH1.
For Research Use Only
Protein Aliases: AC; acid CDase; Acid ceramidase; Acid ceramidase subunit alpha; Acid ceramidase subunit beta; Acylsphingosine deacylase; ASAH; FLJ21558; FLJ22079; N-acylethanolamine hydrolase ASAH1; N-acylsphingosine amidohydrolase; N-acylsphingosine amidohydrolase (acid ceramidase) 1; PHP; PHP32; Putative 32 kDa heart protein
Gene Aliases: AC; ACDase; ASAH; ASAH1; HSD-33; HSD33; PHP; PHP32; SMAPME
UniProt ID: (Human) Q13510
Entrez Gene ID: (Human) 427
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.