Invitrogen
Manufacturer Code:PA534831
Catalog # PIPA534831
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 197 and 395 of ASAH1 (Uniprot IDNo.Q13510) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AC; AC ACDase ASAH PHP PHP32 SMAPME acid CDase acid ceramidase acylsphingosine deacylase putative 32 kDa heart protein; acid CDase; Acid ceramidase; Acid ceramidase subunit alpha; Acid ceramidase subunit beta; Acylsphingosine deacylase; N-acylethanolamine hydrolase ASAH1; N-acylsphingosine amidohydrolase; N-acylsphingosine amidohydrolase (acid ceramidase) 1; PHP32; Putative 32 kDa heart protein
Gene Aliases: AC; ACDase; ASAH; ASAH1; HSD-33; HSD33; PHP; PHP32; SMAPME
UniProt ID: (Human) Q13510
Entrez Gene ID: (Human) 427
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.