Invitrogen
Manufacturer Code:PA5109416
Catalog # PIPA5109416
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 140-389 of human ASAH1 (NP_001120977.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2310081N20Rik AC ACDase Acid ceramidase Acid ceramidase subunit alpha Acid ceramidase subunit beta Acylsphingosine deacylase Asah ASAH1 HSD33 HSD-33 N-acylethanolamine hydrolase ASAH1 N-acylsphingosine amidohydrolase PHP PHP32 Putative 32 kDa heart protein SMAPME; AC; ACDase; acid CDase; Acid ceramidase; Acid ceramidase subunit alpha; Acid ceramidase subunit beta; Acylsphingosine deacylase; N-acylethanolamine hydrolase ASAH1; N-acylsphingosine amidohydrolase; N-acylsphingosine amidohydrolase (acid ceramidase) 1; N-acylsphingosine amidohydrolase 1; PHP32; Putative 32 kDa heart protein
Gene Aliases: 2310081N20Rik; AC; ACDase; ASAH; ASAH1; HSD-33; HSD33; PHP; PHP32; SMAPME
UniProt ID: (Human) Q13510, (Mouse) Q9WV54, (Rat) Q6P7S1
Entrez Gene ID: (Human) 427, (Mouse) 11886, (Rat) 84431
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.