Invitrogen
Manufacturer Code:PA5106787
Catalog # PIPA5106787
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human ARX corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: aristaless related homeobox Aristaless-related homeobox aristaless-related homeobox X-linked Arx arx {ECO:0000250 cancer/testis antigen 121 CT121 EIEE1 homeobox protein ARX ISSX MRX29 MRX32 MRX33 MRX36 MRX38 MRX43 MRX54 MRX76 MRX87 MRXS1 PRTS RGD1562672 UniProtKB:Q96QS3}; Aristaless-related homeobox; aristaless-related homeobox, X-linked; arx {ECO:0000250|UniProtKB:Q96QS3}; cancer/testis antigen 121; Homeobox protein ARX
Gene Aliases: ARX; CT121; EIEE1; ISSX; MRX29; MRX32; MRX33; MRX36; MRX38; MRX43; MRX54; MRX76; MRX87; MRXS1; PRTS; RGD1562672
UniProt ID: (Human) Q96QS3, (Mouse) O35085, (Rat) A6YP92
Entrez Gene ID: (Human) 170302, (Mouse) 11878, (Rat) 317268
Molecular Function:
DNA binding protein
helix-turn-helix transcription factor
homeobox transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.