Novus Biologicals
Manufacturer Code:NBP280505
Catalog # NB013578
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide from human Homeobox protein ARX conjugated to blue carrier protein was used as the antigen. The peptide is homologous in rat and mouse. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: aristaless related homeobox Aristaless-related homeobox aristaless-related homeobox X-linked cancer/testis antigen 121 CT121 EIEE1 homeobox protein ARX ISSXmental retardation X-linked 36 mental retardation X-linked 29 mental retardation X-linked 32 mental retardation X-linked 33 mental retardation X-linked 38 mental retardation X-linked 43 mental retardation X-linked 54 mental retardation X-linked 76 mental retardation X-linked 87 MRX29 MRX32 MRX33 MRX36 MRX38 MRX43 MRX54 MRX76 MRX87 MRXS1 PRTS; Aristaless-related homeobox; aristaless-related homeobox, X-linked; cancer/testis antigen 121; Homeobox protein ARX
Gene Aliases: ARX; CT121; EIEE1; ISSX; MRX29; MRX32; MRX33; MRX36; MRX38; MRX43; MRX54; MRX76; MRX87; MRXS1; PRTS
UniProt ID: (Human) Q96QS3
Entrez Gene ID: (Human) 170302
Molecular Function:
DNA binding protein
helix-turn-helix transcription factor
homeobox transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.