Proteintech
Manufacturer Code:207391AP150UL
Catalog # 50-173-3057
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: aristaless related homeobox ARX Homeobox protein ARX ISSX MRX29 MRX32 MRX33 MRX36 MRX38 MRX43 MRX54 MRX76 MRX87 MRXS1 PRTS; Aristaless-related homeobox; aristaless-related homeobox, X-linked; cancer/testis antigen 121; Homeobox protein ARX
Gene Aliases: ARX; CT121; EIEE1; ISSX; MRX29; MRX32; MRX33; MRX36; MRX38; MRX43; MRX54; MRX76; MRX87; MRXS1; PRTS
UniProt ID: (Human) Q96QS3, (Mouse) O35085
Entrez Gene ID: (Human) 170302, (Mouse) 11878
Molecular Function:
DNA binding protein
helix-turn-helix transcription factor
homeobox transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.