Invitrogen
Manufacturer Code:PA524151
Catalog # PIPA524151
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 23-51 amino acids from the N-terminal region of human ARV1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ARV1 fatty acid homeostatsis modulator; ARV1 homolog, fatty acid homeostatsis modulator; hARV1; hARV1 protein ARV1; Protein ARV1
Gene Aliases: 1110067L22Rik; AI461928; ARV1; AW121084; HT035
UniProt ID: (Human) Q9H2C2, (Mouse) Q9D0U9
Entrez Gene ID: (Human) 64801, (Mouse) 68865
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.