Invitrogen
Manufacturer Code:PA5113911
Catalog # PIPA5113911
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human ARV1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ARV1 ARV1 fatty acid homeostatsis modulator ARV1 homolog (S. cerevisiae) ARV1 homolog fatty acid homeostasis modulator ARV1 homolog fatty acid homeostatsis modulator hARV1 HT035 Protein ARV1 RGD1310264; ARV1 fatty acid homeostatsis modulator; ARV1 homolog, fatty acid homeostatsis modulator; hARV1; Protein ARV1
Gene Aliases: ARV1; HT035
UniProt ID: (Human) Q9H2C2
Entrez Gene ID: (Human) 64801
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.