Proteintech
Manufacturer Code:138211AP150UL
Catalog # 50-172-9026
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ARTS1 Fusion Protein Ag4918 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A LAP ALAP Aminopeptidase PILS APPILS ARTS 1 ARTS1 ERAAP ERAAP1 ERAP1 KIAA0525 PILS AP PILSAP; A-LAP; Adipocyte-derived leucine aminopeptidase; Aminopeptidase PILS; aminopeptidase regulator of TNFR1 shedding; ARTS-1; CTD-2260A17.2; Endoplasmic reticulum aminopeptidase 1; endoplasmic reticulum aminopeptidase 1 delta-Exon-11 isoform; endoplasmic reticulum aminopeptidase 1 delta-Exon-13 isoform; endoplasmic reticulum aminopeptidase 1 delta-Exon-14 isoform; endoplasmic reticulum aminopeptidase 1 delta-Exon-15 isoform; endoplasmic reticulum aminopeptidase associated with antigen processing; ER-aminopeptidase 1; leucyl-specific aminopeptidase PILS; PILS-AP; Puromycin-insensitive leucyl-specific aminopeptidase; Type 1 tumor necrosis factor receptor shedding aminopeptidase regulator; VEGF-induced aminopeptidase
Gene Aliases: A-LAP; ALAP; APPILS; ARTS-1; ARTS1; ERAAP; ERAAP1; ERAP1; KIAA0525; PILS-AP; PILSA; PILSAP; UNQ584/PRO1154
UniProt ID: (Human) Q9NZ08, (Rat) Q9JJ22, (Mouse) Q9EQH2
Entrez Gene ID: (Human) 51752, (Rat) 80897, (Mouse) 80898
Molecular Function: hydrolase metalloprotease protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.