Invitrogen
Manufacturer Code:PA519390
Catalog # PIPA519390
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (ELSKFVKDFSGN) corresponding to the internal amino acids of 38231 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Apoptosis-related protein in the TGF-beta signaling pathway; ARTS; Bradeion beta; Brain protein H5; CE5B3 beta; Cell division control-related protein 2; cell division control-related protein 2b; Cerebral protein 7; hCDCREL-2; M-Septin; peanut-like 2 homolog; Peanut-like protein 2; SEPT4 septin 4 ARTS BRADEION CE5B3 H5 MART PNUTL2 SEP4 hCDCREL-2 hucep-7 cell division control-related protein 2 cerebral protein 7 peanut-like 2 septin-M; septin H5; Septin-4; septin-M
Gene Aliases: ARTS; Bh5; BRADEION; C17orf47; CE5B3; H5; hCDCREL-2; hucep-7; MART; OTTMUSG00000001265; PNUTL2; SEP4; SEPT4; SEPTIN4
UniProt ID: (Human) O43236, (Mouse) Q3UZC3
Entrez Gene ID: (Human) 5414, (Mouse) 18952
Molecular Function: G-protein cytoskeletal protein enzyme modulator small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.