Invitrogen
Manufacturer Code:PA5115882
Catalog # PIPA5115882
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human SEPT4 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 4-Sep Apoptosis-related protein in the TGF-beta signaling pathway ARTS Bh5 BRADEION Bradeion beta brain protein H5 C17orf47 CE5B3 CE5B3 beta cell division control-related protein 2 cell division control-related protein 2b Cerebral protein 7 EG3-1RVC EG3RVC expression gene 3 in rat visual cortex expression gene 3-1 in rat visual cortex H5 hCDCREL-2 hucep-7 MART M-Septin OTTMUSG00000001265 peanut-like 2 peanut-like 2 homolog peanut-like protein 2 PNUTL2 SEP4 Sept4 septin 4 septin H5 Septin4 septin-4 septin-M Uncharacterized protein C17orf47; Apoptosis-related protein in the TGF-beta signaling pathway; ARTS; Bradeion beta; Brain protein H5; CE5B3 beta; Cell division control-related protein 2; cell division control-related protein 2b; Cerebral protein 7; expression gene 3 in rat visual cortex; expression gene 3-1 in rat visual cortex; hCDCREL-2; M-Septin; peanut-like 2; peanut-like 2 homolog; Peanut-like protein 2; septin H5; Septin-4; septin-M
Gene Aliases: ARTS; Bh5; BRADEION; C17orf47; CE5B3; EG3-1RVC; EG3RVC; H5; hCDCREL-2; hucep-7; MART; OTTMUSG00000001265; PNUTL2; SEP4; SEPT4; SEPTIN4
UniProt ID: (Human) O43236, (Mouse) Q3UZC3
Entrez Gene ID: (Human) 5414, (Mouse) 18952, (Rat) 287606
Molecular Function: G-protein cytoskeletal protein enzyme modulator small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.