Invitrogen
Manufacturer Code:PA130832
Catalog # PIPA130832
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide KRFLEDTTDDGE corresponding to amino acids 3-14 of Human ARTS. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Apoptosis-related protein in the TGF-beta signaling pathway; ARTS; ARTS BRADEION CE5B3 H5 MART PNUTL2 SEP4 hCDCREL-2 hucep-7; Bradeion beta; Brain protein H5; CE5B3 beta; Cell division control-related protein 2; Cerebral protein 7; hCDCREL-2; Peanut-like protein 2; Septin-4; septin-M
Gene Aliases: ARTS; BRADEION; C17orf47; CE5B3; H5; hCDCREL-2; hucep-7; MART; PNUTL2; SEP4; SEPT4; SEPTIN4
UniProt ID: (Human) O43236
Entrez Gene ID: (Human) 5414
Molecular Function:
G-protein
cytoskeletal protein
enzyme modulator
small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.