Invitrogen
Manufacturer Code:PA577152
Catalog # PIPA577152
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human ART5. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADP-ribosyltransferase C2 and C3 toxin-like 5; ARTC5; Ecto-ADP-ribosyltransferase 5; Ecto-ADP-ribosyltransferase 5 ADP-ribosyltransferase C2 and C3 toxin-like 5 ARTC5 Mono(ADP-ribosyl)transferase 5 NAD(P)(+)--arginine ADP-ribosyltransferase 5 ART5 UNQ575/PRO1137; Mono(ADP-ribosyl)transferase 5; NAD(P)(+)--arginine ADP-ribosyltransferase 5; YAC-2
Gene Aliases: ART5; ARTC5; UNQ575/PRO1137; Yac-2
UniProt ID: (Human) Q96L15, (Mouse) P70352, (Rat) Q5XHY4
Entrez Gene ID: (Human) 116969, (Mouse) 11875, (Rat) 259167
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.