Proteintech
Manufacturer Code:159301AP150UL
Catalog # 50-173-0580
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ART3 Fusion Protein Ag8775 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADP ribosyltransferase 3 ART3 Ecto ADP ribosyltransferase 3 Mono(ADP ribosyl)transferase 3 TMART; ADP-ribosyltransferase C2 and C3 toxin-like 3; ART3 mono(ADP-ribosyl)transferase; ARTC3; Ecto-ADP-ribosyltransferase 3; Mono(ADP-ribosyl)transferase 3; mono-ADP-ribosyltransferase; mono-ADP-ribosyltransferase 3; NAD(P)(+)--arginine ADP-ribosyltransferase 3
Gene Aliases: 4930569O04Rik; ART3; ARTC3; TMART
UniProt ID: (Human) Q13508, (Mouse) Q8R2G4
Entrez Gene ID: (Human) 419, (Mouse) 109979
Molecular Function: glycosyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.