Invitrogen
Manufacturer Code:MA528162
Catalog # PIMA528162
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | Nika102 |
Immunogen | pMECD8LF-ART22 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: ADP-ribosyltransferase C2 and C3 toxin-like 2; ARTC2; Mono(ADP-ribosyl)transferase 2B; NAD(+) glycohydrolase; T-cell differentiation marker Rt6 homolog 2; T-cell ecto-ADP-ribosyltransferase 2; T-cell mono(ADP-ribosyl)transferase 2; T-cell NAD(P)(+)--arginine ADP-ribosyltransferase 2
Gene Aliases: Art; ART2.2; Art2b; ARTC2; Ly92b; Rt-6; Rt6; Rt6-2; Rt6.2
UniProt ID: (Mouse) O35975
Entrez Gene ID: (Mouse) 11872
Molecular Function: glycosyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.