Novus Biologicals
Manufacturer Code:NBP291995100UL
Catalog # NB025883
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 23-295 of human ART1 (NP_004305.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADP-ribosyltransferase 1 ADP-ribosyltransferase 2 CD296 GPI-linked NAD(P)(+)--arginine ADP-ribosyltransferase 1 MGC133217 mono(ADP-ribosyl)transferase 1 RT6; ADP-ribosyltransferase 2; ADP-ribosyltransferase C2 and C3 toxin-like 1; ARTC1; CD296; GPI-linked NAD(P)(+)--arginine ADP-ribosyltransferase 1; Mono(ADP-ribosyl)transferase 1
Gene Aliases: ART1; ART2; ARTC1; CD296; RT6
UniProt ID: (Human) P52961
Entrez Gene ID: (Human) 417
Molecular Function: glycosyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.