Proteintech
Manufacturer Code:190611AP150UL
Catalog # 50-173-2329
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ARSA Fusion Protein Ag13569 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ARSA arylsulfatase A ASA Cerebroside sulfatase MLD; Arylsulfatase A; Arylsulfatase A component B; Arylsulfatase A component C; ASA; Cerebroside-sulfatase
Gene Aliases: ARSA; As-2; AS-A; As2; ASA; AW212749; MLD; TISP73
UniProt ID: (Human) P15289, (Mouse) P50428
Entrez Gene ID: (Human) 410, (Rat) 315222, (Mouse) 11883
Molecular Function: hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.