Invitrogen
Manufacturer Code:PA5109414
Catalog # PIPA5109414
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-270 of human ARRDC3 (NP_065852.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI450344 Arrdc3 Arrestin domain-containing protein 3 Kiaa1376 Liver regeneration-related protein LRRG00048 LRRGT00048 mKIAA1376 TBP-2-like inducible membrane protein TLIMP; alpha-arrestin 3; Arrestin domain-containing protein 3; Liver regeneration-related protein LRRG00048; TBP-2-like inducible membrane protein; thioredoxin-binding protein-2-like inducible membrane; TLIMP
Gene Aliases: ARRDC3; KIAA1376; LRRGT00048; TLIMP
UniProt ID: (Human) Q96B67, (Rat) Q6TXF1
Entrez Gene ID: (Human) 57561, (Rat) 309945
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.