Novus Biologicals
Manufacturer Code:NBP292055100UL
Catalog # NB026002
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-270 of human ARRDC3 (NP_065852.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alpha-arrestin 3; arrestin domain containing 3 arrestin domain-containing protein 3 KIAA1376TLIMPTBP-2-like inducible membrane protein thioredoxin-binding protein-2-like inducible membrane; Arrestin domain-containing protein 3; TBP-2-like inducible membrane protein; thioredoxin-binding protein-2-like inducible membrane; TLIMP
Gene Aliases: ARRDC3; KIAA1376; TLIMP
UniProt ID: (Human) Q96B67
Entrez Gene ID: (Human) 57561
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.