Abnova
Manufacturer Code:PAB17489
Catalog # 89-119-149
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to C-terminus of human ARRDC3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of ARRDC3.
For Research Use Only
Protein Aliases: alpha-arrestin 3; Arrestin domain-containing protein 3; KIAA1376; TBP-2-like inducible membrane protein; thioredoxin-binding protein-2-like inducible membrane; TLIMP
Gene Aliases: ARRDC3; KIAA1376; TLIMP
UniProt ID: (Human) Q96B67
Entrez Gene ID: (Human) 57561
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.