Invitrogen
Manufacturer Code:PA599592
Catalog # PIPA599592
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human Arrestin-C corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ARR3 Arr4 arrestin 3 arrestin 3 retinal (X-arrestin) arrestin 3 retinal arrestin 3 retinal (X-arrestin) arrestin 4 Arrestin C ArrestinC arrestin-C ARRX CAR Carfl cArr C-arrestin cone arrestin Retinal cone arrestin-3 X-arrestin; arrestin 3 retinal (X-arrestin); arrestin 4; Arrestin-C; C-arrestin; cArr; Cone arrestin; Retinal cone arrestin-3; X-arrestin
Gene Aliases: ARR3; Arr4; ARRX; CAR; Carfl; Carr
UniProt ID: (Human) P36575, (Mouse) Q9EQP6
Entrez Gene ID: (Human) 407, (Mouse) 170735
Molecular Function:
enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.