Novus Biologicals
Manufacturer Code:NBP292277100UL
Catalog # NB026445
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-100 of human ARPC5L (NP_112240.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: actin related protein 2/3 complex subunit 5-like; actin related protein 2/3 complex subunit 5-like actin-related protein 2/3 complex subunit 5-like protein ARC16-2MGC3038 Arp2/3 complex 16 kDa subunit 2; Actin-related protein 2/3 complex subunit 5-like protein; ARC16-2; Arp2/3 complex 16 kDa subunit 2
Gene Aliases: ARC16-2; ARPC5L
UniProt ID: (Human) Q9BPX5
Entrez Gene ID: (Human) 81873
Molecular Function: actin family cytoskeletal protein cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.