Proteintech
Manufacturer Code:220251AP150UL
Catalog # 50-173-3691
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ARPC5L Fusion Protein Ag16881 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: actin related protein 2/3 complex subunit 5-like; Actin-related protein 2/3 complex subunit 5-like protein; ARC16 2 ARPC5L; ARC16-2; Arp2/3 complex 16 kDa subunit 2
Gene Aliases: 2010015J01Rik; AI852867; ARC16-2; ARPC5L; AW555592; AW742746
UniProt ID: (Human) Q9BPX5, (Rat) A1L108, (Mouse) Q9D898
Entrez Gene ID: (Human) 81873, (Rat) 296710, (Mouse) 74192
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.