Abnova
Manufacturer Code:H00081873K
Catalog # 89-203-734
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human ARPC5L |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human ARPC5L peptide using ARM Technology.
For Research Use Only
Protein Aliases: actin related protein 2/3 complex subunit 5-like; Actin-related protein 2/3 complex subunit 5-like protein; ARC16-2; ARC16-2 MGC3038; Arp2/3 complex 16 kDa subunit 2
Gene Aliases: ARC16-2; ARPC5L
UniProt ID: (Human) Q9BPX5
Entrez Gene ID: (Human) 81873
Molecular Function: actin family cytoskeletal protein cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.