Proteintech
Manufacturer Code:109301AP150UL
Catalog # 50-172-6723
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ARPC4 Fusion Protein Ag1366 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: actin related protein 2/3 complex subunit 4, 20kDa; actin related protein 2/3 complex, subunit 4, 20kDa; Actin-related protein 2/3 complex subunit 4; ARC20 Arp2/3 complex 20 kDa subunit ARPC4 p20 Arc; Arp2/3 complex 20 kDa subunit; arp2/3 protein complex subunit p20; p20-ARC
Gene Aliases: 20kDa; 5330419I20Rik; AI327076; ARC20; ARPC4; P20-ARC
UniProt ID: (Human) P59998, (Mouse) P59999
Entrez Gene ID: (Human) 10093, (Mouse) 68089, (Rat) 297518
Molecular Function: actin family cytoskeletal protein cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.