Proteintech
Manufacturer Code:138221AP150UL
Catalog # 50-172-9027
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ARP3/ARP3B Fusion Protein Ag4922 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Actin like protein 3 Actin related protein 3 ACTR3 ACTR3B Arp11 ARP3 Arp4; Actin-like protein 3; Actin-related protein 3; actin-related protein 3 homolog (yest); ARP3 actin-related protein 3 homolog
Gene Aliases: 1200003A09Rik; ACTR3; ARP3
UniProt ID: (Human) P61158, (Rat) Q4V7C7, (Mouse) Q99JY9
Entrez Gene ID: (Human) 10096, (Rat) 81732, (Mouse) 74117
Molecular Function:
actin and actin related protein
actin family cytoskeletal protein
cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.