Abnova
Manufacturer Code:H00010097B01
Catalog # 89-006-893
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | ACTR2 (AAH36253.1 1 a.a. ~ 339 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human ACTR2 protein.
For Research Use Only
Protein Aliases: Actin-like protein 2; Actin-related protein 2; ARP2; ARP2 actin-related protein 2 homolog
Gene Aliases: ACTR2; ARP2
UniProt ID: (Human) P61160
Entrez Gene ID: (Human) 10097
Molecular Function:
actin and actin related protein
actin family cytoskeletal protein
cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.