Invitrogen
Manufacturer Code:PA517311
Catalog # PIPA517311
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to the sequence of human ARP2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Actin-like protein 14D; Actin-like protein 2; actin-like protein 2 actin-related protein 2; Actin-related protein 14 Da; Actin-related protein 14D; Actin-related protein 2; Arp14D; ARP2 actin-related protein 2 homolog; ARP2 actin-related protein 2 homolog (yeast); Arp2-PA; Arp2-PB; Arp2-PC; CG9901-PA; CG9901-PB; CG9901-PC; LD18955p
Gene Aliases: 4921510D23Rik; AA409782; Actr14D; ACTR2; ARP14D; ARP2; CG9901; D6Ertd746e; Dmel\CG9901; Dmel_CG9901; I79_003843
UniProt ID: (Human) P61160, (Rat) Q5M7U6, (Mouse) P61161, (Fruit fly) P45888
Entrez Gene ID: (Human) 10097, (Rat) 289820, (Mouse) 66713, (Chinese hamster) 100752591, (Fruit fly) 32623
Molecular Function:
actin and actin related protein
actin family cytoskeletal protein
cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.