Proteintech
Manufacturer Code:109221AP150UL
Catalog # 50-172-6716
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ARP2 Fusion Protein Ag1359 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Actin like protein 2 Actin related protein 2 ARP2; Actin-like protein 2; Actin-related protein 2; ARP2 actin-related protein 2 homolog
Gene Aliases: 4921510D23Rik; AA409782; ACTR2; ARP2; D6Ertd746e
UniProt ID: (Human) P61160, (Rat) Q5M7U6, (Mouse) P61161
Entrez Gene ID: (Human) 10097, (Rat) 289820, (Mouse) 66713
Molecular Function:
actin and actin related protein
actin family cytoskeletal protein
cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.