OriGene
Manufacturer Code:TA501174
Catalog # 50-167-2318
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1C11 |
Immunogen | Full length human recombit protein of human ARNTL produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ARNT-like protein 1, brain and muscle; Aryl hydrocarbon receptor nuclear translocator-like protein 1; basic-helix-loop-helix-PAS orphan MOP3; Basic-helix-loop-helix-PAS protein MOP3; bHLH-PAS protein JAP3; bHLHe5; BMAL1 BMAL1c JAP3 MOP3 PASD3 TIC bHLHe5; Brain and muscle ARNT-like 1; Class E basic helix-loop-helix protein 5; Member of PAS protein 3; member of PAS superfamily 3; PAS domain-containing protein 3; testis tissue sperm-binding protein Li 50e
Gene Aliases: ARNTL; BHLHE5; BMAL1; BMAL1c; JAP3; MOP3; PASD3; TIC
UniProt ID: (Human) O00327
Entrez Gene ID: (Human) 406
Molecular Function: basic helix-loop-helix transcription factor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.