Invitrogen
Manufacturer Code:PA572632
Catalog # PIPA572632
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Zebrafish |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 475-501 amino acids from the Central region of DANRE arnt2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Ah receptor nuclear translocator; Arnt; ARNT protein 2; ARNT2A; ARNT2b; ARNT2c; Aryl hydrocarbon receptor nuclear translocator 2; Aryl hydrocarbon receptor nuclear translocator 2 ARNT protein 2 Class E basic helix-loop-helix protein 1 bHLHe1 ARNT 2 BHLHE1 KIAA0307; unm t30256; unm_t30256
Gene Aliases: Arnt; arnt2; arnt2a; arnt2X; chunp6921; zfARNT2; zfARNT2a; zfARNT2b; zfARNT2c
UniProt ID: (Zebrafish) Q9DG12
Entrez Gene ID: (Zebrafish) 64277
Molecular Function:
basic helix-loop-helix transcription factor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.