Invitrogen
Manufacturer Code:PA571950
Catalog # PIPA571950
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 249-278 amino acids from the Central region of human ARNT2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ARNT protein 2; Aryl hydrocarbon receptor nuclear translocator 2; Aryl hydrocarbon receptor nuclear translocator 2 ARNT protein 2 Class E basic helix-loop-helix protein 1 bHLHe1 ARNT 2 BHLHE1 KIAA0307; aryl-hydrocarbon receptor nuclear translocator 2; bHLHe1; Class E basic helix-loop-helix protein 1
Gene Aliases: ARNT2; BHLHE1; KIAA0307; WEDAS
UniProt ID: (Human) Q9HBZ2
Entrez Gene ID: (Human) 9915
Molecular Function: basic helix-loop-helix transcription factor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.