Invitrogen
Manufacturer Code:PA530338
Catalog # PIPA530338
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 129 and 371 of Human ARNT2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ARNT protein 2; Aryl hydrocarbon receptor nuclear translocator 2; aryl-hydrocarbon receptor nuclear translocator 2; bHLHe1; bHLHe1 ARNT protein 2 aryl hydrocarbon receptor nuclear translocator 2 class E basic helix-loop-helix protein 1; Class E basic helix-loop-helix protein 1
Gene Aliases: ARNT2; BHLHE1; Hif-2b; KIAA0307; mKIAA0307; WEDAS
UniProt ID: (Human) Q9HBZ2, (Mouse) Q61324
Entrez Gene ID: (Human) 9915, (Mouse) 11864
Molecular Function: basic helix-loop-helix transcription factor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.