Novus Biologicals
Manufacturer Code:NBP215469
Catalog # NBP215469
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human ARNT2. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ARNT protein 2; ARNT protein 2 aryl-hydrocarbon receptor nuclear translocator 2 bHLHe1BHLHE1 Class E basic helix-loop-helix protein 1 KIAA0307aryl hydrocarbon receptor nuclear translocator 2; Aryl hydrocarbon receptor nuclear translocator 2; aryl-hydrocarbon receptor nuclear translocator 2; bHLHe1; Class E basic helix-loop-helix protein 1
Gene Aliases: ARNT2; BHLHE1; KIAA0307; WEDAS
UniProt ID: (Human) Q9HBZ2
Entrez Gene ID: (Human) 9915
Molecular Function: basic helix-loop-helix transcription factor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.