Novus Biologicals
Manufacturer Code:NB300525
Catalog # NB300525
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2B10 |
Immunogen | Synthetic peptide corresponding to residues C N(771) S Y N N E E F P D L T M F P P F S E(789) of human ARNT. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ARNT protein; ARNT protein aryl hydrocarbon receptor nuclear translocator BHLHE2 HIF-1 Beta HIF1B HIF-1-beta HIF1-beta HIF1BETA hypoxia-inducible factor 1 beta subunit Hypoxia-inducible factor 1-beta nuclear translocator; Aryl hydrocarbon receptor nuclear translocator; bHLHe2; Class E basic helix-loop-helix protein 2; Dioxin receptor, nuclear translocator; HIF-1-beta; hypoxia-inducible factor 1, beta subunit; Hypoxia-inducible factor 1-beta
Gene Aliases: ARNT; BHLHE2; HIF-1-beta; HIF-1beta; HIF1-beta; HIF1B; HIF1BETA; TANGO
UniProt ID: (Human) P27540
Entrez Gene ID: (Human) 405
Molecular Function:
basic helix-loop-helix transcription factor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.