Invitrogen
Manufacturer Code:PA520061
Catalog # PIPA520061
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A peptide corresponding to 15 amino acids near the C-terminus of human ARMER. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADP-ribosylation factor GTPase 6 interacting protein 1; ADP-ribosylation factor like GTPase 6 interacting protein 1; ADP-ribosylation factor-like 6 interacting protein 1; ADP-ribosylation factor-like protein 6-interacting protein 1; ADP-ribosylation-like factor 6 interacting protein; aip-1; AIP1 ARL6IP ARMER ADP-ribosylation factor-like protein 6-interacting protein 1 ARL-6-interacting protein 1 aip-1 apoptotic regulator in the membrane of the endoplasmic reticulum; Apoptotic regulator in the membrane of the endoplasmic reticulum; ARL-6-interacting protein 1; Protein TBX2
Gene Aliases: Aip-1; AIP-6; AIP1; AL022945; ARL6IP; ARL6IP1; ARMER; AU042858; C85138; KIAA0069; mKIAA0069; SPG61
UniProt ID: (Human) Q15041, (Mouse) Q9JKW0
Entrez Gene ID: (Human) 23204, (Mouse) 54208
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.