Abnova
Manufacturer Code:H00055130M02A
Catalog # 89-107-010
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 5F1 |
Immunogen | ARMC4 (NP_060546945 a.a. &bsim 1044 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant ARMC4.
For Research Use Only
Protein Aliases: Armadillo repeat-containing protein 4; DKFZp434P1735; FLJ10376; FLJ10817; FLJ32798; Outer dynein arm-docking complex subunit 2; RP11-691I13.1; testis tissue sperm-binding protein Li 74n
Gene Aliases: ARMC4; CILD23; ODAD2
UniProt ID: (Human) Q5T2S8
Entrez Gene ID: (Human) 55130
Molecular Function:
cell adhesion molecule
cytoskeletal protein
signaling molecule
storage protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.