Novus Biologicals
Manufacturer Code:NBP2844600.1ML
Catalog # NB032388
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the N-terminal region of ARMC4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: armadillo repeat containing 4 armadillo repeat-containing protein 4 DKFZP434P1735 FLJ10376 FLJ10817 FLJ32798; Armadillo repeat-containing protein 4; Outer dynein arm-docking complex subunit 2; testis tissue sperm-binding protein Li 74n
Gene Aliases: ARMC4; CILD23; ODAD2
UniProt ID: (Human) Q5T2S8
Entrez Gene ID: (Human) 55130
Molecular Function: cell adhesion molecule cytoskeletal protein signaling molecule storage protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.