Abnova
Manufacturer Code:PAB12918
Catalog # 89-116-187
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to C-terminus 15 amino acids of human ARL6IP1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for three months. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of ARL6IP1.
For Research Use Only
Protein Aliases: ADP-ribosylation factor GTPase 6 interacting protein 1; ADP-ribosylation factor like GTPase 6 interacting protein 1; ADP-ribosylation factor-like 6 interacting protein 1; ADP-ribosylation factor-like protein 6-interacting protein 1; aip-1; AIP1; Apoptotic regulator in the membrane of the endoplasmic reticulum; ARL-6-interacting protein 1; ARL6IP; ARMER; KIAA0069
Gene Aliases: AIP1; ARL6IP; ARL6IP1; ARMER; KIAA0069; SPG61
UniProt ID: (Human) Q15041
Entrez Gene ID: (Human) 23204
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.