Invitrogen
Manufacturer Code:PA5100129
Catalog # PIPA5100129
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human BBS3 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1110018H24Rik 2210411E14Rik ADP ribosylation factor like GTPase 6 ADP-ribosylation factor-like 6 ADP-ribosylation factor-like protein 6 ADP-ribosylation-like 6 ADP-ribosylation-like factor homolog ARL6 Arl6 Bardet-Biedl syndrome 3 protein BBS3 RP55; ADP-ribosylation factor-like protein 6; ADP-ribosylation-like 6; ADP-ribosylation-like factor homolog ARL6; Bardet-Biedl syndrome 3 protein
Gene Aliases: 1110018H24Rik; 2210411E14Rik; ARL6; BBS3; RP55
UniProt ID: (Human) Q9H0F7, (Mouse) O88848
Entrez Gene ID: (Human) 84100, (Rat) 363760, (Mouse) 56297
Molecular Function: G-protein enzyme modulator small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.