Novus Biologicals
Manufacturer Code:NBP29262820UL
Catalog # NB027127
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-186 of human ARL6 (NP_115522.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADP-ribosylation factor-like 6 ADP-ribosylation factor-like protein 6 Bardet-Biedl syndrome 3 protein BBS3 MGC32934 RP55; ADP-ribosylation factor-like protein 6; Bardet-Biedl syndrome 3 protein
Gene Aliases: ARL6; BBS3; RP55
UniProt ID: (Human) Q9H0F7
Entrez Gene ID: (Human) 84100
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.