OriGene
Manufacturer Code:TA800297
Catalog # 50-167-8238
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1A10 |
Immunogen | Full length human recombit protein of human ARL2BP produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ADP-ribosylation factor like 2 binding protein; ADP-ribosylation factor like GTPase 2 binding protein; ADP-ribosylation factor-like 2 binding protein; ADP-ribosylation factor-like protein 2-binding protein; Arf-like 2 binding protein BART1; ARF-like 2-binding protein; ARL2-binding protein; BART BART1 RP66; Binder of ARF2 protein 1; binder of Arl Two; binder of Arl2; retinitis pigmentosa 66 (autosomal recessive)
Gene Aliases: ARL2BP; BART; BART1; RP66
UniProt ID: (Human) Q9Y2Y0
Entrez Gene ID: (Human) 23568
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.