Invitrogen
Manufacturer Code:PA527091
Catalog # PIPA527091
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to a region within amino acids 123 and 184 of ARL2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADP-ribosylation factor like GTPase 2; ADP-ribosylation factor-like 2; ADP-ribosylation factor-like protein 2; ARFL2 ADP-ribosylation factor-like protein 2
Gene Aliases: ARFL2; ARL2
UniProt ID: (Human) P36404
Entrez Gene ID: (Human) 402
Molecular Function:
G-protein
enzyme modulator
small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.