Invitrogen
Manufacturer Code:PA5104310
Catalog # PIPA5104310
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human ARL13B corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A530097K21Rik A930014M17Rik ADP ribosylation factor like GTPase 13B ADP-ribosylation factor like GTPase 13B ADP-ribosylation factor-like 13B ADP-ribosylation factor-like 2-like 1 ADP-ribosylation factor-like protein 13B ADP-ribosylation factor-like protein 2-like 1 Arl13b Arl2l1 ARL2-like protein 1 C530009C10Rik hnn JBTS8 Protein hennin; ADP-ribosylation factor like GTPase 13B; ADP-ribosylation factor-like 13B; ADP-ribosylation factor-like 2-like 1; ADP-ribosylation factor-like protein 13B; ADP-ribosylation factor-like protein 2-like 1; ARL2-like protein 1; Protein hennin
Gene Aliases: A530097K21Rik; A930014M17Rik; ARL13B; ARL2L1; C530009C10Rik; hnn; JBTS8
UniProt ID: (Human) Q3SXY8, (Mouse) Q640N2
Entrez Gene ID: (Dog) 487934, (Human) 200894, (Rat) 304037, (Mouse) 68146
Molecular Function: G-protein enzyme modulator small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.