Novus Biologicals
Manufacturer Code:NBP214312
Catalog # NBP214312
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids: ATAKGIQGEYPEDVAPTVGFSKINLRQGKFEVTIFDLGGGIRIRGIWKNY YAESYGVIFVVDSSDEERMEETKEAMSEMLR |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADP-ribosylation factor like GTPase 13B; ADP-ribosylation factor-like 13B; ADP-ribosylation factor-like 13B ADP-ribosylation factor-like 2-like 1 ADP-ribosylation factor-like protein 13B ADP-ribosylation factor-like protein 2-like 1 ARL2L1MGC120611 ARL2-like protein 1 DKFZp686E2075 DKFZp686L2472 DKFZp761H079 JBTS8DKFZp686M2074 MGC120612; ADP-ribosylation factor-like 2-like 1; ADP-ribosylation factor-like protein 13B; ADP-ribosylation factor-like protein 2-like 1; ARL2-like protein 1
Gene Aliases: ARL13B; ARL2L1; JBTS8
UniProt ID: (Human) Q3SXY8
Entrez Gene ID: (Human) 200894
Molecular Function:
G-protein
enzyme modulator
small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.