Novus Biologicals
Manufacturer Code:NBP183622
Catalog # NBP183622
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:TSKYPSRDMYRESENSSFPSHRHQEKLHVNYLTSLHLQDKKSAAAEAPTDDQPTDLSLPKNPHKPTGKVLGLAHSTTGPQESKGISQFQVLGSQSRDCHPKACRVSPMTMSGPKKYPESLSRSGKPHHVRLENFRKME |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ARID domain-containing protein 5B; ARID domain-containing protein 5B AT rich interactive domain 5B (MRF1-like) DESRTAT-rich interactive domain-containing protein 5B FLJ21150 Modulator recognition factor 2 modulator recognition factor 2 (MRF2) MRF-2 MRF2MRF1-like protein; AT rich interactive domain 5B (MRF1-like); AT-rich interactive domain 5B (MRF1-like); AT-rich interactive domain-containing protein 5B; Modulator recognition factor 2; modulator recognition factor 2 (MRF2); MRF-2; MRF1-like protein
Gene Aliases: ARID5B; DESRT; MRF-2; MRF2
UniProt ID: (Human) Q14865
Entrez Gene ID: (Human) 84159
Molecular Function: transcription cofactor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.